missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinonitrile, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.
Solubility
Slightly soluble in methanol and water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.
Solubility
Slightly soluble in methanol and water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
Specifications
Specifications
Melting Point | 52°C to 56°C |
Boiling Point | 265°C to 267°C |
Flash Point | 132°C (269°F) |
Odor | Odorless |
Quantity | 100 g |
UN Number | UN3439 |
Beilstein | 1098380 |
Merck Index | 14,8872 |
Solubility Information | Slightly soluble in methanol and water. |
Formula Weight | 80.09 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'