missing translation for 'onlineSavingsMsg'
Learn More

Succinonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11341807
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.

Solubility
Slightly soluble in methanol and water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-61-2
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00001949
InChI Key IAHFWCOBPZCAEA-UHFFFAOYSA-N
Synonym succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil
PubChem CID 8062
IUPAC Name butanedinitrile
SMILES N#CCCC#N

Specifications

Melting Point 52°C to 56°C
Boiling Point 265°C to 267°C
Flash Point 132°C (269°F)
Odor Odorless
Quantity 500 g
UN Number UN3439
Beilstein 1098380
Merck Index 14,8872
Solubility Information Slightly soluble in methanol and water.
Formula Weight 80.09
Percent Purity 98%
Chemical Name or Material Succinonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'