missing translation for 'onlineSavingsMsg'
Learn More

Succinyl chloride, 96%, Thermo Scientific Chemicals

£53.37 - £204.00

Chemical Identifiers

CAS 543-20-4
Molecular Formula C4H4Cl2O2
Molecular Weight (g/mol) 154.974
MDL Number MFCD00000749
InChI Key IRXBNHGNHKNOJI-UHFFFAOYSA-N
Synonym succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate
PubChem CID 10970
IUPAC Name butanedioyl dichloride
SMILES C(CC(=O)Cl)C(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11448317
View Documents
Thermo Scientific Alfa Aesar
A18389.18
50 g
£53.37
50g
Please to purchase this item. Need a web account? Register with us today!
11458317
View Documents
Thermo Scientific Alfa Aesar
A18389.30
250 g
£204.00
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is employed in the synthesis of biseugenyl succinate and bis(4-maleimidephenyl) succinate. It is also used as used as an important raw material and intermediate in pharmaceuticals, organic synthesis, agrochemicals and dye stuffs. It is also applied as hydrolytic reagent for the determination of water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of biseugenyl succinate and bis(4-maleimidephenyl) succinate. It is also used as used as an important raw material and intermediate in pharmaceuticals, organic synthesis, agrochemicals and dye stuffs. It is also applied as hydrolytic reagent for the determination of water.

Solubility
Reacts violently with water.

Notes
Moisture Sensitive. Store away from oxidizing agents, bases, alcohols and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Chemical Identifiers

543-20-4
154.974
IRXBNHGNHKNOJI-UHFFFAOYSA-N
10970
C(CC(=O)Cl)C(=O)Cl
C4H4Cl2O2
MFCD00000749
succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate
butanedioyl dichloride

Specifications

543-20-4
1.407
76°C (168°F)
C4H4Cl2O2
MFCD00000749
507064
14,8873
Reacts violently with water.
C(CC(=O)Cl)C(=O)Cl
154.974
154.98
Succinyl chloride
16°C to 17°C
190°C
Acrid
1.471
UN3265
Moisture sensitive
succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate
IRXBNHGNHKNOJI-UHFFFAOYSA-N
butanedioyl dichloride
10970
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 208-838-0

missing translation for 'rtecsNumber' : WN4900000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only