missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sucrose, ACS

Product Code. 11367968
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11367968

Brand: Thermo Scientific Chemicals 036508.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 57-50-1 | C12H22O11 | 342.30 g/mol

Sucrose is widely utilized in medications to carry a more pleasant taste to indigestible chemicals. It is used in many medical dosage forms such as chewable tablets, syrups, tablets, or glues. Sucrose is mainly used in foods nowadays. The fatty acid esters of sucrose show useful application in transdermal therapeutic systems. Sucrose-gradient centrifugation also plays an important role in the selective isolation of nocardia spp. from soil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 185°C to 187°C
Density 1.58 g/mL
Quantity 250 g
Beilstein 90825
Merck Index 14,8881
Solubility Information Soluble in water; Slightly soluble in alcohol,methanol; Insoluble in diethyl ether.
Formula Weight 342.3
Grade ACS
Physical Form Crystalline
Chemical Name or Material Sucrose, For analysis ACS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.