Learn More
Sucrose Brix/RI Standards, Paragon Scientific™
Paragon ISO 17025 and ISO 17034 certified brix standards are for the calibration and verification of all types of refractometer e.g handheld, Abbe and high accuracy digital instruments. Typically used within the food and beverage and pharmaceutical industries.
£52.00 - £81.25
Product Code | Brand | Concentration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Price | Quantity & Availability | |||||
16080400
|
Paragon Scientific
SS05 |
5.00 Brix |
£54.00
15mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090400
|
Paragon Scientific
SS075 |
7.50 Brix |
£52.00
15mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16000410
|
Paragon Scientific
SS10 |
10.00 Brix |
£81.25
15mL |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040410
|
Paragon Scientific
SS125 |
12.50 Brix |
£52.00
15mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050410
|
Paragon Scientific
SS15 |
15.00 Brix |
£52.00
15mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16070410
|
Paragon Scientific
SS20 |
20.00 Brix |
£52.00
15mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16080410
|
Paragon Scientific
SS25 |
25.00 Brix |
£62.35
15mL |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090410
|
Paragon Scientific
SS30 |
30.00 Brix |
£52.00
15mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010420
|
Paragon Scientific
SS35 |
35 Brix |
£52.00
15mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16020420
|
Paragon Scientific
SS40 |
40.00 Brix |
£52.00
15mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16030420
|
Paragon Scientific
SS45 |
45.00 Brix |
£52.00
15mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040420
|
Paragon Scientific
SS50 |
50.00 Brix |
£62.35
15mL |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050420
|
Paragon Scientific
SS55 |
55 Brix |
£52.00
15mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060420
|
Paragon Scientific
SS60 |
60.00 Brix |
£81.15
15mL |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Primary level refractive index standards traceable to National Standards
- Manufactured in accordance with ICUMSA Methods
- Supplied in tamper evident packs with 6 weeks shelf life
Nominal values quoted