missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Specifications
Packaging | Poly Bottle |
Quantity | 500 g |
Specific Gravity | 1L = 1.59kg |
Formula Weight | 342.3 |
Grade | BAKER ANALYZED™ ACS Reagent |
Physical Form | Crystals |
Chemical Name or Material | Sucrose |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'