missing translation for 'onlineSavingsMsg'
Learn More

Sucrose, Crystal, Multi-Compendial, N.F., J.T. Baker™

Catalog Number 15588234
227.05 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
57-50-1
C12H22O11
342.30
MFCD00006626
CZMRCDWAGMRECN-PWPRYFECNA-N
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
5988
CHEBI:17992
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
This item is not returnable. View return policy
57-50-1
C12H22O11
342.30
MFCD00006626
CZMRCDWAGMRECN-PWPRYFECNA-N
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
5988
CHEBI:17992
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1L = 1.59kg
Formula Weight 342.3
Grade Multi-Compendial/NF
Physical Form Crystals
Chemical Name or Material Sucrose
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'