missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose, Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Loss on Drying | Max. 0.03 % |
Packaging | Plastic Bottle |
Quantity | 250 g |
Percent Purity | 99.0 - 100.0 % |
Grade | Reagent Pharmacopeia Organics |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'