missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose octaacetate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | >78.0°C |
Color | Cream-White to White |
Density | 1.2800g/mL |
Boiling Point | 250.0°C (1.0 mmHg) |
Ignition Residue | 0.1% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass Bottle |
Quantity | 5 g |
Merck Index | 15,9013 |
Show More |
Safety and Handling
Product Identifier
- Sucrose octaacetate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'