missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan Black B

Product Code. 15404679
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Histochemical stain for chromosomes

Histochemical stain for chromosomesSudan Black B is used as a biological stain for lipids. It is a basic black dye utilized in staining triglycerides and lipids off some lipoproteins. Further, it is used in the fingerprint enhancement as well as for detecting fats. In addition to this, it is used in the study of hematological disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Histochemical stain for chromosomesSudan Black B is used as a biological stain for lipids. It is a basic black dye utilized in staining triglycerides and lipids off some lipoproteins. Further, it is used in the fingerprint enhancement as well as for detecting fats. In addition to this, it is used in the study of hematological disorders.

Solubility
Soluble in oils, fats, warm petrolatum, paraffin, phenol, ethanol, acetone, benzene, toluene and hydrocarbon. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4197-25-5
Molecular Formula C29H24N6
Molecular Weight (g/mol) 456.553
MDL Number MFCD00006919
InChI Key YCUVUDODLRLVIC-UHFFFAOYSA-N
Synonym C.I. 26150; Solvent Black 3
PubChem CID 61336
IUPAC Name (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene
SMILES CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C

Specifications

Melting Point 118°C to 126°C
Beilstein 723248
Merck Index 14,8885
Solubility Information Soluble in oils,fats,warm petrolatum,paraffin,phenol,ethanol,acetone,benzene,toluene and hydrocarbon. Insoluble in water.
Formula Weight 456.55
Color Black to Brown
Quantity 100 g
Physical Form Powder
Chemical Name or Material Sudan Black B
compliance-icons
Product Identifier
  • Sudan Black B
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sudan Black B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.