missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sudan Black B, high purity biological stain
£52.16 - £141.71
Chemical Identifiers
CAS | 4197-25-5 |
---|---|
Molecular Formula | C29H24N6 |
Molecular Weight (g/mol) | 456.553 |
MDL Number | MFCD00006919 |
InChI Key | YCUVUDODLRLVIC-UHFFFAOYSA-N |
Synonym | C.I. 26150, Fat Black HB, Solvent Black 3 |
PubChem CID | 61336 |
IUPAC Name | (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene |
SMILES | CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10616212
|
Thermo Scientific Chemicals
190160250 |
25 g | Glass bottle |
£52.16
25g |
In Stock
Log in to see stock. |
||||
10475125
|
Thermo Scientific Chemicals
190161000 |
100 g | Glass bottle |
£141.71
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4197-25-5 | |
456.553 | |
YCUVUDODLRLVIC-UHFFFAOYSA-N | |
61336 | |
CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C |
C29H24N6 | |
MFCD00006919 | |
C.I. 26150, Fat Black HB, Solvent Black 3 | |
(2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene |
Specifications
120.0°C to 124.0°C | |
C29H24N6 | |
C.I. 26150, Fat Black HB, Solvent Black 3 | |
YCUVUDODLRLVIC-UHFFFAOYSA-N | |
CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C | |
456.553 | |
456.54 | |
Complies | |
Fine Crystalline Powder |
4197-25-5 | |
MFCD00006919 | |
Solubility in water: insoluble. | |
Authentic | |
(2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene | |
61336 | |
High Purity | |
Black to Brown | |
Sudan Black B, high purity biological stain |
Safety and Handling
Product Identifier
- Sudan Black B
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title