missing translation for 'onlineSavingsMsg'
Learn More

Sudan orange G, pure, Thermo Scientific Chemicals

Product Code. 10350494
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-85-6
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002275
InChI Key PULNESMFDLBKAZ-SDNWHVSQSA-N
Synonym C.I. 11920, 4-(Phenylazo)resorcinol, Solvent orange 1
PubChem CID 5921655
IUPAC Name (4Z)-3-hydroxy-4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1

Specifications

Infrared Spectrum Authentic
Formula Weight 214.22
Grade Extra Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Orange to Red
Quantity 100 g
Physical Form Powder
Chemical Name or Material Sudan orange G, Pure
compliance-icons
Product Identifier
  • Sudan Orange G
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'