missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfacetamide, 98%, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Type | Sulfacetamide |
Melting Point | 182.0°C |
Infrared Spectrum | Authentic |
Loss on Drying | 0.5% max. (105°C, 2 hrs) |
Quantity | 500 g |
Formula Weight | 214.24 |
Percent Purity | ≥97.5% (HPLC) |
Chemical Name or Material | Sulfacetamide |
Safety and Handling
Product Identifier
- Sulfacetamide
- Warning
- Germ cell mutagenicity Category 2
- H341-Suspected of causing genetic defects.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'