missing translation for 'onlineSavingsMsg'
Learn More

Sulfachloropyridazine sodium, 97%, Thermo Scientific™

Product Code. 15502333
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 23282-55-5
Molecular Formula C10H8ClN4NaO2S
Molecular Weight (g/mol) 306.7
InChI Key GHQJXOBLWKFNAD-UHFFFAOYSA-N
PubChem CID 87077948
IUPAC Name 4-amino-N-(6-chloropyridazin-3-yl)benzenesulfonamide;sodium
SMILES C1=CC(=CC=C1N)S(=O)(=O)NC2=NN=C(C=C2)Cl.[Na]

Specifications

Melting Point 262.0°C to 270.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 306.7
Percent Purity 97%
Chemical Name or Material Sulfachloropyridazine sodium
compliance-icons
Product Identifier
  • Sulphachloropyridazine sodium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium N-(6-chloropyridazin-3-yl)sulphanilamidate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'