missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamic Acid, 98%, Reagent grade, Honeywell™
Click to view available options
Quantity:
1 kg
3 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Appearance | Powder |
Color | White |
Melting Point | 209°C |
Density | 2.1g/cm3 |
Packaging | Poly Bottle |
Quantity | 3 kg |
Linear Formula | NH2SO3H |
UN Number | UN2967 |
Formula Weight | 97.09g/mol |
Percent Purity | 98% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'