missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamic Acid, Certified AR for Analysis, Fisher Chemical
Brand: Fisher Chemical S/8641/53
87.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
500g, Plastic powder jar

Specifications
5329-14-6 | |
White | |
Plastic powder jar | |
11603 | |
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid | |
NS(=O)(=O)O | |
97.088 | |
CHEBI:9330 | |
Solid |
200°C | |
1.2 | |
H3NO3S | |
500 g | |
IIACRCGMVDHOTQ-UHFFFAOYSA-N | |
sulfamic acid | |
5987 | |
97.09g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sulfamic acid
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sulfamic Acid, Certified AR for Analysis, Fisher Chemical > 500g, Plastic powder jar
Spot an opportunity for improvement?Share a Content Correction