Learn More
Sulfamide, 99%, Thermo Scientific Chemicals
£26.53 - £243.21
Chemical Identifiers
CAS | 7803-58-9 |
---|---|
Molecular Formula | H4N2O2S |
Molecular Weight (g/mol) | 96.10 |
MDL Number | MFCD00011606 |
InChI Key | NVBFHJWHLNUMCV-UHFFFAOYSA-N |
Synonym | sulfuric diamide, sulfamamide, sulfuryl amide, sulfonyl diamide, sulfuryl diamide, sulfamoylamine, sulphamide, sulphuric diamide, unii-vs7tzw634v, imidosulfamic acid |
PubChem CID | 82267 |
ChEBI | CHEBI:29368 |
SMILES | NS(N)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11422968
|
Thermo Scientific Alfa Aesar
|
2.5 g |
£26.53
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11432968
|
Thermo Scientific Alfa Aesar
|
10 g |
£64.32
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442968
|
Thermo Scientific Alfa Aesar
|
50 g |
£243.21
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.
Solubility
Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol
Notes
Moisture Sensitive. Store away from oxidizing agents, water/humidity. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
7803-58-9 | |
96.10 | |
NVBFHJWHLNUMCV-UHFFFAOYSA-N | |
82267 | |
NS(N)(=O)=O |
H4N2O2S | |
MFCD00011606 | |
sulfuric diamide, sulfamamide, sulfuryl amide, sulfonyl diamide, sulfuryl diamide, sulfamoylamine, sulphamide, sulphuric diamide, unii-vs7tzw634v, imidosulfamic acid | |
CHEBI:29368 |
Specifications
7803-58-9 | |
1.61 | |
MFCD00011606 | |
14,8922 | |
Soluble in water, hot alcohol, acetone; Sparingly soluble in cold alcohol | |
NS(N)(=O)=O | |
96.10 | |
CHEBI:29368 | |
99% |
91°C to 92°C | |
H4N2O2S | |
Moisture sensitive | |
sulfuric diamide, sulfamamide, sulfuryl amide, sulfonyl diamide, sulfuryl diamide, sulfamoylamine, sulphamide, sulphuric diamide, unii-vs7tzw634v, imidosulfamic acid | |
NVBFHJWHLNUMCV-UHFFFAOYSA-N | |
sulfamide | |
82267 | |
96.11 | |
Sulfamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
missing translation for 'einecsNumber' : 232-262-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only