missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sulfanilamide 98%

Product Code. 10091350 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10091350

Brand: Thermo Scientific Chemicals 132855000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sulfanilamide, CAS # 63-74-1, is an organic sulfur compound that shows antibacterial activities.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sulfanilamide is an organic sulfur compound with antibacterial properties that are structurally similar to the p-aminobenzoic acid (PABA)
  • It also has a competitive inhibitory capacity against dihydropteroate synthetases blocking the synthesis of folic acid
  • Sulfanilamide competes with PABA for the bacterial enzyme dihydropteroate synthase preventing, the incorporation of PABA into dihydrofolic acid, an immediate precursor of folic acid. This culminates in the inhibition of the bacterial folic acid synthesis and also the de novo synthesis of purines and pyrimidines, resulting in cell growth arrest and cell death.

Applications

  • This compound shows antibacterial activity against different species, including gram-positive, gram-negative, and Chlamydia
  • In chemical reactions, this compound can act as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution
  • Sulfanilamide shows inhibitory activity against the enzyme, carbonic anhydrase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-74-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00007939
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym 4-Aminobenzenesulfonamide
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)N

Specifications

Type Sulfanilamide
Physical Form Crystalline Powder
Melting Point 163.0°C to 167.0°C
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (1 g, 105°C)
Packaging Plastic Bottle
Color White
Quantity 500 g
Assay Percent Range 98%
Linear Formula 4-(H2N)C6H4SO2NH2
Beilstein 14, 698
Merck Index 15, 9057
Solubility Information Solubility in water: 7.5g/L (20°C).
Formula Weight 172.2
Percent Purity 98%
Chemical Name or Material Sulfanilamide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Sulfanilamide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.