Learn More
Sulfanilic acid, 99%, Thermo Scientific Chemicals
£23.80 - £692.00
Chemical Identifiers
CAS | 121-57-3 |
---|---|
Molecular Formula | C6H7NO3S |
Molecular Weight (g/mol) | 173.186 |
MDL Number | MFCD00007886 |
InChI Key | HVBSAKJJOYLTQU-UHFFFAOYSA-N |
Synonym | sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid |
PubChem CID | 8479 |
ChEBI | CHEBI:27500 |
IUPAC Name | 4-aminobenzenesulfonic acid |
SMILES | C1=CC(=CC=C1N)S(=O)(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10347610
|
Thermo Scientific Acros
150711000 |
100 g | Plastic bottle |
£23.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10705221
|
Thermo Scientific Acros
150710010 |
1 kg | Plastic bottle |
£105.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10772494
|
Thermo Scientific Acros
150710025 |
2.5 kg | Plastic bottle |
£210.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13438368
|
Thermo Scientific Acros
150710100 |
10 kg | Plastic drum |
£692.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
121-57-3 | |
173.186 | |
HVBSAKJJOYLTQU-UHFFFAOYSA-N | |
8479 | |
4-aminobenzenesulfonic acid |
C6H7NO3S | |
MFCD00007886 | |
sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid | |
CHEBI:27500 | |
C1=CC(=CC=C1N)S(=O)(=O)O |
Specifications
121-57-3 | |
99% | |
(H2N)C6H4SO3H | |
sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid | |
C1=CC(=CC=C1N)S(=O)(=O)O | |
173.186 | |
CHEBI:27500 | |
≥98.5% (Acid) | |
Sulfanilic acid |
Authentic | |
C6H7NO3S | |
MFCD00007886 | |
HVBSAKJJOYLTQU-UHFFFAOYSA-N | |
4-aminobenzenesulfonic acid | |
8479 | |
173.19 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.
GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Sulfanilic acid
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only