missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilic acid, ACS reagent, Thermo Scientific Chemicals

Product Code. 10455471
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-57-3
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.186
MDL Number MFCD00007886
InChI Key HVBSAKJJOYLTQU-UHFFFAOYSA-N
Synonym sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid
PubChem CID 8479
ChEBI CHEBI:27500
IUPAC Name 4-aminobenzenesulfonic acid
SMILES C1=CC(=CC=C1N)S(=O)(=O)O

Specifications

Color White
Ignition Residue 0.01% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula (H2N)C6H4SO3H
Quantity 100 g
Beilstein 14, 695
Merck Index 15, 9058
Solubility Information 0.02% max. Insoluble Matter (in Na2CO3-solution)
Formula Weight 173.19
Percent Purity 98.0 to 102.0%
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material Sulfanilic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'