missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Sulfo-NHS-LC-Biotin (Biotin-X, SSE, 6-((Biotinoyl)Amino)Hexanoic Acid, Sulfosuccinimidyl Ester, Sodium Salt)
Sulfo-NHS-LC-Biotin (Biotin-X, SSE, 6-((Biotinoyl)Amino)Hexanoic Acid, Sulfosuccinimidyl Ester, Sodium Salt)
Brand: Molecular Probes™ B6353
Code : 50
Additional Details : Weight : 0.00525kg
Description
Biotin-X, SSE reacts efficiently and reproducibly with amines. The biotin is well separated from the point of attachment by a spacer consisting of a seven-atom aminohexanoyl group (X). This spacer enhances the accessibility of biotin to the relatively deep biotin-binding sites of avidin and streptavidin. The sulfonate group on the succinimidyl ester increases the water solubility of the reactive moiety.Specifications
Sulfo-NHS-LC-Biotin, 25mg, Off-white, 556.58 Molecular Weight, Biotin-X Dye, Amine Chemical Reactivity, –5 to –30°C Storage, Used For Labeling Chemistry, Protein Labeling and Crosslinking | |
Solid | |
25 mg | |
556.58 | |
Biotin-X |
Solid | |
Labeling chemistry | |
Sulfo-NHS-LC-Biotin | |
-5°C to -30°C | |
Off-White |
For Research Use Only. Not for use in diagnostic procedures.