Learn More
Thermo Scientific Chemicals Sulfobromophthalein sodium salt hydrate
Description
Sulfobromophthalein sodium salt hydrate is a reagent for the quantitative determination of proteins, a dye for liver diagnostics in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Quantity | 1 g |
Sensitivity | Moisture sensitive |
Merck Index | 14,8954 |
Solubility Information | Soluble in water. |
Formula Weight | 837.98 (Anhydrous) |
Physical Form | Powder |
Chemical Name or Material | Sulfobromophthalein sodium salt hydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N07L081 | Certificate of Analysis | 10/02/2025 | 043878.14,043878.03,043878.06 |
Z31K002 | Certificate of Analysis | 06/01/2025 | 043878.03,043878.14,043878.06 |
T18K014 | Certificate of Analysis | 02/08/2024 | 043878.03,043878.14,043878.06 |
U04J012 | Certificate of Analysis | 24/10/2023 | 043878.03,043878.14,043878.06 |
Q14J032 | Certificate of Analysis | 26/04/2023 | 043878.03 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.