missing translation for 'onlineSavingsMsg'
Learn More
Sulfolane, 99%, Thermo Scientific Chemicals Product Code.: 11405543

Sulfolane, 99%, Thermo Scientific Chemicals

Product Code. 11405543
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11405543

Brand: Thermo Scientific Alfa Aesar A13466.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Miscible with water, acetone, toluene

Sulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.

Solubility
Miscible with alcohol, water, acetone, toluene and dilute mineral acids. Slightly miscible with octanes, olefins and naphthenes.

Notes
Miscible with water, acetone, tolueneIncompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-33-0
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00005484
InChI Key HXJUTPCZVOIRIF-UHFFFAOYSA-N
Synonym sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfoneShow MoreShow Less
PubChem CID 31347
ChEBI CHEBI:74794
SMILES O=S1(=O)CCCC1

Specifications

Melting Point 25°C to 28°C
Density 1.261
Boiling Point 285°C
Flash Point 166°C (330°F)
Assay Percent Range 99%
Odor Weak, Characteristic
Refractive Index 1.4825
Quantity 250 g
Beilstein 107765
Sensitivity Hygroscopic
Merck Index 14,8955
Solubility Information Miscible with alcohol,water,acetone,toluene and dilute mineral acids. Slightly miscible with octanes,olefins and naphthenes.
IUPAC Name 1λ⁶-thiolane-1,1-dione
Formula Weight 120.17
Percent Purity 99%
Chemical Name or Material Sulfolane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP31G035 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog NumberA13466.0I,A13466.0B,A13466.30
Lot NumberY19F055 Certificate TypeCertificate of Analysis Date18/03/2025 Catalog NumberA13466.0I,A13466.0B,A13466.30
Lot NumberT10E033 Certificate TypeCertificate of Analysis Date18/03/2025 Catalog NumberA13466.0I,A13466.0B,A13466.30
Lot NumberN04L018 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog NumberA13466.0I,A13466.0B,A13466.30
Lot NumberU05F038 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog NumberA13466.0I,A13466.0B,A13466.30
compliance-icons compliance-icons
Product Identifier
  • Sulfolane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Tetramethylene sulfone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfolane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.