missing translation for 'onlineSavingsMsg'
Learn More

Sulforhodamine 101, Tocris Bioscience™

Product Code. 30002029
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Red fluorescent dye; selective astrocyte marker

Sulforhodamine 101 is a fluorescent dye (excitation maximum ∼ 586 nm; emission maximum ∼ 606). Selective astrocyte marker.

Specifications

CAS 60311-02-6
Molecular Formula C31H29ClN2O6S2
MDL Number MFCD00012407
Quantity 50 mg
Synonym sulforhodamine 101, sulforhodamine 101; sulforhodamine 640, unii-fx0es3271v, texas red, sulforhodamine 101 acid chloride, 2-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-pyrido 3,2,1-ij quinolizino 1',9':6,7,8 chromeno 2,3-f quinolin-4-ium-9-yl-5-sulfobenzenesulfonate, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9$l^ 5 ,23-diazaheptacyclo 17.7.1.1^ 5,9 .0^ 2,17 .0^ 4,15 .0^ 23,27 .0^ 13,28 octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9??,23-diazaheptacyclo 17.7.1.1?,?.0 2 , 1 ?.0?, 1 ?.0 2 3 , 2 ?.0 1 3 , 2 ? octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9lambda,23-diazaheptacyclo 17.7.1.1,.0 2 , 1 .0, 1 .0 2 3 , 2 .0 1 3 , 2 octacosa-1,4,9 28 ,13,15,17,19 27-heptaen-9-ylium, 9-2,4-disulfophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium inner salt, hydrogen 9-2,4-disulphonatophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium
InChI Key MPLHNVLQVRSVEE-UHFFFAOYSA-N
SMILES [O-]S(=O)(=O)C1=CC(=CC=C1C1=C2C=C3CCCN4CCCC(C2=[O+]C2=C5CCCN6CCCC(C=C12)=C56)=C34)S(Cl)(=O)=O
IUPAC Name 16-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3λ⁴-oxa-9,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-3-ylium
Molecular Weight (g/mol) 625.15
PubChem CID 122180
ChEBI CHEBI:52274
Formula Weight 606.71
Chemical Name or Material Sulforhodamine 101
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'