Learn More
Chemical Identifiers
CAS | 7704-34-9 |
---|---|
Molecular Formula | S |
Molecular Weight (g/mol) | 32.06 |
MDL Number | MFCD00085316 |
InChI Key | NINIDFKCEFEMDL-UHFFFAOYSA-N |
Synonym | polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot |
PubChem CID | 5362487 |
ChEBI | CHEBI:26833 |
IUPAC Name | sulfur |
SMILES | [S] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11924061
|
Thermo Scientific Acros
201225000 |
500 g | Plastic bottle |
£32.48
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606242
|
Thermo Scientific Acros
201220025 |
2.5 kg | Plastic bottle |
£46.43
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914061
|
Thermo Scientific Acros
201220100 |
10 kg | Plastic drum |
£145.73
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
7704-34-9 | |
32.06 | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
5362487 | |
sulfur |
S | |
MFCD00085316 | |
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot | |
CHEBI:26833 | |
[S] |
Specifications
445.0°C | |
7704-34-9 | |
Powder | |
S | |
01,1118; 03,273; 05,632; 06,556; 14,291; 15,297 | |
15,9099 | |
Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol and ether, soluble in carbon disulfide, benzene, (about 2.4%, at 30°C, much more at higher temps.),toluene,, acetone (2.65% at 25°C), chloroform (1.5% at 18°C) | |
[S] | |
32.06 | |
CHEBI:26833 | |
≥99.5% |
114.0°C | |
Yellow | |
99.5% min | |
MFCD00085316 | |
168°C | |
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
sulfur | |
5362487 | |
32.05 | |
Sulfur |
Safety and Handling
GHS H Statement
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and w
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-722-6
- Sulfur
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.