missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, 99.999%, (trace metal basis)

Product Code. p-221781 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10401441

Brand: Thermo Scientific Acros 199930100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7704-34-9 | S | 32.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Boiling Point 445.0°C
Melting Point 114.0°C
Color Yellow
Physical Form Pieces
Quantity 10 g
Assay Percent Range 99.999%
Fieser 01,1118; 03,273; 05,632; 06,556; 14,291; 15,297
Flash Point 168°C
Merck Index 15, 9099
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol and ether, soluble in carbon disulfide, benzene, (about 2.4%, at 30°C, much more at higher temps.),toluene,, acetone (2.65% at 25°C), chloroform (1.5% at 18°C)
Formula Weight 32.05
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Density 2.36
Chemical Name or Material Sulfur, 99.999%
Show More Show Less
compliance-icons
Product Identifier
  • Sulphur
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sulphur

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.