missing translation for 'onlineSavingsMsg'
Learn More
Sulfur, flowers, 99%, Thermo Scientific Chemicals Product Code.: 15493147

Sulfur, flowers, 99%, Thermo Scientific Chemicals

Product Code. 15493147
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 15493147

Brand: Thermo Scientific Alfa Aesar A17260.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S

Specifications

Boiling Point 444°C
Melting Point 117°C to 120°C
Physical Form Solid
Quantity 500 g
Percent Purity 99%

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251408 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA17260.22,A17260.0E,A17260.36
Lot Number10249173 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog NumberA17260.22,A17260.0E,A17260.36
Lot Number10229929 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17260.22,A17260.0E,A17260.36
Lot Number10217923 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17260.22,A17260.0E,A17260.36
Lot Number10228022 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17260.22,A17260.0E,A17260.36
compliance-icons
Product Identifier
  • Sulphur, flowers
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sulfur
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfur, flowers, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.