Learn More
Sulfur Oil Standard in No. 2 Diesel Fuel, 1000μg/g, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep SDFS-1000-Y
Code : 04
Additional Details : CAS Number : 68476-34-6 Weight : 0.14000kg
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
S | 1000μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Sulfur Oil Standard in #2 Diesel Fuel | |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
1000μg/g | |
#2 Diesel Fuel |
For Diesel Fuel Analysis | |
100 mL | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Sulphur |
Product intended for laboratory use only