missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur Oil Standard in No. 2 Diesel Fuel, 200μg/g, SPEX CertiPrep™
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
Packaging:
Triple-leached LDPE bottle
Beschreibung
Beinhaltet
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
S | 200μg/g |
Zertifizierungen
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spezifikation
CAS | 544-40-1, 68476-34-6 |
Packaging | Triple-leached LDPE bottle |
Quantity | 100 mL |
CAS Max % | 99.98, 0.02 |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Molecular Formula | C8H18S |
MDL Number | MFCD00009468 |
InChI Key | HTIRHQRTDBPHNZ-UHFFFAOYSA-N |
SMILES | CCCCSCCCC |
Mehr anzeigen |
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'