missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur Oil Standards for Diesel Fuel Analysis, SPEX CertiPrep™
Click to view available options
Concentration:
10 μg/g
100 μg/g
1000 μg/g
200 μg/g
25 μg/g
300 μg/g
5 μg/g
50 μg/g
75 μg/g
750 μg/g
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
S | 5μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Concentration | 5 μg/g |
Packaging | Triple-leached LDPE bottle |
CAS Min % | 0.0, 99.99 |
Quantity | 100 mL |
CAS Max % | 0.0, 99.99 |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Solvent or Matrix | Base Oil 20 |
Physical Form | Solid |
Chemical Name or Material | Sulfur Oil Standard in Base Oil |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Sulfur Oil Standard in Base Oil 20
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Mineral Oil
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'