missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur, precipitated, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 1 kg |
UN Number | 1350 |
Flash Point | 207°C (404.6°F) |
Solubility Information | In accordance with carbon disulfide, 1g/5mL |
Formula Weight | 32.07g/mol |
Percent Purity | 99.5 to 100.5% |
Grade | puriss. |
Ignition Point | 450°F |
Residue after Ignition | ≤0.2% (as SO4) |
Vapor Pressure | 1mmHg (183.8°C), 10mmHg (246°C) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction