missing translation for 'onlineSavingsMsg'
Learn More
Sulfur, Reagent grade, Purified by refining, Honeywell™ Product Code.: 15641980

Sulfur, Reagent grade, Purified by refining, Honeywell™

Product Code. 15641980 Shop All Honeywell Research Chemicals Products
500 g, Poly Bottle
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15641980

Brand: Honeywell Research Chemicals 215198500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, powder, purified by refining, -100 mesh particle size

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Boiling Point 445°C
Melting Point 120°C
Color Yellow
Physical Form Powder
Quantity 500 g
UN Number UN1350
Flash Point 207°C (404.6°F)
Formula Weight 32.07g/mol
Grade Reagent
Ignition Point 450°F
Packaging Poly Bottle
Vapor Pressure 1mmHg (183.8°C), 10mmHg (246°C)
Density 2.07g/cu.cm; Vapor Density: 8.9 (vs air)
Chemical Name or Material Sulfur, Purified by refining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfur, Reagent grade, Purified by refining, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.