missing translation for 'onlineSavingsMsg'
Learn More

Sulfur trioxide-pyridine complex, 98%, active SO3 ca 48-50%, Thermo Scientific Chemicals

Product Code. 11464863
Click to view available options
Quantity:
10 g
50 g
250 g
1000 g

Sulfur trioxide-pyridine complex is used for sulfations of alcohols, sulfonations, deoxygenations and other reductions, oxidations with dimethyl sulfoxide. It is also used as a source of sulfur trioxide especially in the preparation of sulfate esters. Further, it is used as an activating electrophile in a Parikh-Doering oxidation. In addition to this, it is also used in the preparation and anticoagulation test of sulfated silk fibroin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfur trioxide-pyridine complex is used for sulfations of alcohols, sulfonations, deoxygenations and other reductions, oxidations with dimethyl sulfoxide. It is also used as a source of sulfur trioxide especially in the preparation of sulfate esters. Further, it is used as an activating electrophile in a Parikh-Doering oxidation. In addition to this, it is also used in the preparation and anticoagulation test of sulfated silk fibroin.

Solubility
Soluble in polar organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26412-87-3
Molecular Formula C5H5NO3S
Molecular Weight (g/mol) 159.16
MDL Number MFCD00012437
InChI Key UDYFLDICVHJSOY-UHFFFAOYSA-N
Synonym pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide
PubChem CID 168533
SMILES O=S(=O)=O.C1=CC=NC=C1

Specifications

Melting Point ∼155°C to 165°C
Quantity 1000 g
UN Number UN3261
Beilstein 3704116
Sensitivity Moisture sensitive
Solubility Information Soluble in polar organic solvents.
IUPAC Name pyridine;sulfur trioxide
Formula Weight 159.16
Percent Purity 98%
Chemical Name or Material Sulfur trioxide-pyridine complex, Active SO3 ≈48 to 50%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'