missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur trioxide pyridine complex, technical, 48-50% active SO3
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
| Melting Point | 155.0°C |
| Color | White to Beige |
| Infrared Spectrum | Authentic |
| Packaging | Plastic bottle |
| Linear Formula | SO3·C5H5N |
| Quantity | 2.5 kg |
| Beilstein | 20,19 |
| Fieser | 01,1127; 02,393; 03,275; 04,473; 08,391 |
| Solubility Information | Solubility in water: reacts. Other solubilities: soluble in conc. h2so4,hcl and hclo4,soluble in dmf |
| IUPAC Name | pyridine;sulfur trioxide |
| Show More |
Safety and Handling
Product Identifier
- Sulfur trioxide pyridine complex, 48-50% active SO3
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction