Learn More
Sulfur trioxide trimethylamine complex, 95%, Thermo Scientific Chemicals
£25.80 - £211.00
Chemical Identifiers
CAS | 3162-58-1 |
---|---|
Molecular Formula | C3H9NO3S |
Molecular Weight (g/mol) | 139.17 |
MDL Number | MFCD00012421 |
InChI Key | DXASQZJWWGZNSF-UHFFFAOYSA-N |
Synonym | sulfur trioxide trimethylamine complex, sulfur trioxide trimethylamine, sulfur trioxide-trimethylamine complex, sulphur trioxide trimethylamine 1:1, sulfur trioxide-trimethylamine, trimethylamine sulfur trioxide, sulfur trioxide; trimethylamine, 1n1&1&&so3 complex, trimethylamine, compd. with sulfur trioxide 1:1, trimethylamine, compound with sulphur trioxide |
PubChem CID | 222852 |
IUPAC Name | N,N-dimethylmethanamine;sulfur trioxide |
SMILES | CN(C)C.O=S(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10055823
|
Thermo Scientific Acros
159350250 |
25 g | Glass bottle |
£25.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10687124
|
Thermo Scientific Acros
159351000 |
100 g | Glass bottle |
£62.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10484765
|
Thermo Scientific Acros
159355000 |
500 g | Glass bottle |
£211.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
3162-58-1 | |
139.17 | |
DXASQZJWWGZNSF-UHFFFAOYSA-N | |
222852 | |
CN(C)C.O=S(=O)=O |
C3H9NO3S | |
MFCD00012421 | |
sulfur trioxide trimethylamine complex, sulfur trioxide trimethylamine, sulfur trioxide-trimethylamine complex, sulphur trioxide trimethylamine 1:1, sulfur trioxide-trimethylamine, trimethylamine sulfur trioxide, sulfur trioxide; trimethylamine, 1n1&1&&so3 complex, trimethylamine, compd. with sulfur trioxide 1:1, trimethylamine, compound with sulphur trioxide | |
N,N-dimethylmethanamine;sulfur trioxide |
Specifications
3162-58-1 | |
White | |
95% | |
(CH3)3N·SO3 | |
01,1128; 08,466 | |
Solubility in water: insoluble. Other solubilities: soluble in hot water, ethanol | |
CN(C)C.O=S(=O)=O | |
139.17 | |
139.17 | |
Powder |
232.0°C | |
Authentic | |
C3H9NO3S | |
MFCD00012421 | |
sulfur trioxide trimethylamine complex, sulfur trioxide trimethylamine, sulfur trioxide-trimethylamine complex, sulphur trioxide trimethylamine 1:1, sulfur trioxide-trimethylamine, trimethylamine sulfur trioxide, sulfur trioxide; trimethylamine, 1n1&1&&so3 complex, trimethylamine, compd. with sulfur trioxide 1:1, trimethylamine, compound with sulphur trioxide | |
DXASQZJWWGZNSF-UHFFFAOYSA-N | |
N,N-dimethylmethanamine;sulfur trioxide | |
222852 | |
95% | |
Sulfur trioxide trimethylamine complex |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 221-614-7
- Sulfur trioxide trimethylamine complex
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only