missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.01M, Reagecon™

Product Code. 11593022
Click to view available options
Quantity:
5 L
Packaging:
5L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Concentration 0.01M
Packaging 5L
Normality 0.2N
Quantity 5 L
compliance-icons
Product Identifier
  • Analytical Volumetric Solution Sulphuric Acid 0.01M (0.02N)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'