missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 124230025

 View more versions of this product

Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10473055

  • £79.19 / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sulfuric acid
0.1N (0.05M) Standard Solution
100.0°C
1.3g/mL
2.5 L
H2SO4
MFCD00064589
15, 9104
Solubility in water: miscible.
OS(O)(=O)=O
98.07
CHEBI:26836
Pure
Colorless
7664-93-9, 7732-18-5
0.0998 to 0.1002N (20°C)
Plastic Bottle
H2O4S
Liquid
01,470; 05,633; 06,558; 07,347; 09,441
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.07
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sulfuric acid, 0.1N (0.05M) standard solution

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only