missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
20 L
Description
Sulfuric acid, 0.5N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 L |
Color | Colorless to Yellow |
Formula Weight | 98.08 |
Concentration or Composition (by Analyte or Components) | Sulfuric acid: 5%; Water: 95% |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Solubility | Miscible with water |
Physical Form | Liquid |
Chemical Name or Material | Sulfuric acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'