missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 1N (0.5M) standard solution

Product Code. 10607753 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Plastic bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10607753

Brand: Thermo Scientific Acros 124240025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfuric acid
Color Colorless
Name Note 1N (0.5M) Standard Solution
CAS 7664-93-9, 7732-18-5
Boiling Point 100.0°C
Concentration 0.998 to 1.002N (20°C)
Density 1.05g/mL
Packaging Plastic bottle
Quantity 2.5 L
Molecular Formula H2O4S
Linear Formula H2SO4
Physical Form Liquid
MDL Number MFCD00064589
Fieser 01,470; 05,633; 06,558; 07,347; 09,441
Merck Index 15, 9104
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.07
Grade Pure
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid, 1N standard solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.