Learn More
Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
£46.63 - £86.83
Specifications
Chemical Name or Material | Sulfuric acid |
---|---|
Color | Colorless |
Name Note | 1N (0.5M) Standard Solution |
CAS | 7664-93-9, 7732-18-5 |
Concentration | 0.998 to 1.002N (20°C) |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10041200
|
Thermo Scientific Acros
124240010 |
1 L | Plastic bottle |
£46.63
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10607753
|
Thermo Scientific Acros
124240025 |
2.5 L | Plastic bottle |
£86.83
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionSpecifications
Sulfuric acid | |
1N (0.5M) Standard Solution | |
0.998 to 1.002N (20°C) | |
Plastic bottle | |
H2SO4 | |
MFCD00064589 | |
15, 9104 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.07 |
Colorless | |
7664-93-9, 7732-18-5 | |
1.05g/mL | |
H2O4S | |
Liquid | |
01,470; 05,633; 06,558; 07,347; 09,441 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
Pure |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Sulfuric acid, 1N standard solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : Sulfuric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.