missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 4M, Chem-Lab

Ideal for titrations in laboratory applications. Sulfuric Acid, 4M, Chem-Lab is a solution of 392.32g H2SO4 per liter of water for use in volumetric analysis. Also known as battery acid, which is used in lead-acid batteries.

Chemical Identifiers

CAS 7664-93-9, 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
View More Specs

Description

Description

Sulfuric acid is a highly corrosive, strong mineral acid that shows different properties depending upon its concentration. It is a pungent-ethereal, colorless to slightly yellow viscous liquid. Sulfuric acid is a diprotic acid that is hygroscopic. Concentrated sulfuric acid has strong dehydrating and oxidizing properties. Dilute sulfuric acid is a good electrolyte.
Specifications

Chemical Identifiers

7664-93-9, 7732-18-5
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O

Specifications

<1
7664-93-9, 7732-18-5
0°C
4mol/L
Odorless
8N (±0.016, 20°C)
Liquid
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08g/mol
Colorless
392.32g H2SO4 per 1L H2O
100°C
1.23g/mL
Polyethylene
H2O4S
MFCD00064589
Soluble in Water
OS(O)(=O)=O
98.07
CHEBI:26836
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep container tightly closed.

Product Identifier
  • Sulfuric acid 4 mol/l

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid

SDS
Documents

Documents

Product Certifications