missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 7.00, Chem-Lab
Designed for pH measurement and maintenance in pH-neutral samples. Buffer Solution, pH 7.00, Chem-Lab is a phosphate solution with a pH level accurate to ±0.02. The solution is traceable to Standard Reference Materials (SRM) from NIST.
Specifications
Melting Point | 0°C |
---|---|
Boiling Point | 100°C |
Color | Colorless |
Density | 1.01g/mL |
For Use With (Application) | Laboratory use, pH measurements |
Description
pH Levels ±0.02:- pH 7.13 — 0°C
- pH 7.05 — 10°C
- pH 7.00 — 20°C
- pH 6.99 — 25°C
- pH 6.98 — 30°C
- pH 6.97 — 40°C
- pH 6.96 — 50°C
- pH 6.96 — 60°C
- pH 6.97 — 70°C
- pH 6.98 — 80°C
- pH 7.00 — 90°C
Specifications
0°C | |
Colorless | |
Laboratory use, pH measurements | |
Liquid | |
Phosphate | |
Yes |
100°C | |
1.01g/mL | |
7.00 ±0.02 | |
pH Buffer | |
Odorless | |
Soluble in water |