missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric acid 62%, d=1.52, for the determination of fats in cheese, Fisher Chemical™ Product Code.: 16370464

Sulfuric acid 62%, d=1.52, for the determination of fats in cheese, Fisher Chemical™

Product Code. 16370464
5 L
Click to view available options
Quantity:
1 L
5 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 16370464

Brand: Fisher Chemical S/9370/21

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Suitable for the determination of fats in cheese.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9, 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Density 1.52
Packaging HDPE Plastic Bottle
Assay Percent Range 61.5-62.5%
Quantity 5 L
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric acid 62%, d=1.52, for the determination of fats in cheese, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.