missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid 93-98%, Trace Metal™, for trace metal analysis, Fisher Chemical

Product Code. 11395820 Shop All Fisher Chemical Products
Click to view available options
:
500mL
This item is not returnable. View return policy

Product Code. 11395820

Brand: Fisher Chemical undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

pH 0.3
Color Brown to Colorless
CAS 7664-93-9
Melting Point 10°C
Boiling Point 290°C
Vapor Pressure <0.001mmHg at 20°C
Quantity 500 mL
Molecular Formula H2O4S
Physical Form Liquid
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
Formula Weight 98.08g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid, 94-98%
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sulfuric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.