missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid 93-98%, Trace Metal™, for trace metal analysis, Fisher Chemical
476.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
2.5L
Specifications
pH | 1 |
Color | Colorless |
CAS | 7664-93-9 |
Melting Point | 10°C |
Boiling Point | 290°C |
Vapor Pressure | 1mmHg 146 |
Quantity | 2.5 L |
Molecular Formula | H2O4S |
Viscosity | 21mPa at 25°C |
Physical Form | Liquid |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'