missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 95.0 to 98.0%, N.F. - F.C.C., J.T. Baker™
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Specifications
Specifications
Packaging | Glass Bottle |
Specific Gravity | 1L = 1.84kg |
Quantity | 2.5 L |
Formula Weight | 98.08 |
Percent Purity | 95.0 to 98.0% |
Grade | FCC/NF |
Chemical Name or Material | Sulfuric Acid |
Safety and Handling
Signal Word
- Danger
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction