missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid 95-98% d=1.83, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™

£245.00 - £503.00

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10620714
View Documents
Fisher Chemical
S/9231/PB08
500 mL Plastic Coated Glass bottle
£257.00
500mL
Please to purchase this item. Need a web account? Register with us today!
10466463
View Documents
Fisher Chemical
S/9231/PB15
1 L HDPE plastic bottle
£245.00
1 L
Please to purchase this item. Need a web account? Register with us today!
10707263
View Documents
Fisher Chemical
S/9231/PB17
2.5 L HDPE plastic bottle
£503.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sulfuric Acid 95-98% d=1.83, Primar Plus™, for Trace Metal Analysis

Specifications

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O

Specifications

1
7664-93-9
290°C
H2O4S
Liquid
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
Colorless
10°C
1mmHg 146
21mPa at 25°C
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.07g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sulfuric acid

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications