missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, ACS Reagent, 95.0 to 98.0%, Honeywell Fluka™
ACS Reagent, 95.0-98.0%
£25.15 - £377.41
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15622780
|
Honeywell-Fluka
258105-100ML |
100 mL | Glass Bottle |
£41.48
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15662780
|
Honeywell-Fluka
258105-4X100ML |
4 x 100 mL | Glass Bottle | N/A | |||||
15692780
|
Honeywell-Fluka
258105-500ML-PC |
500 mL | Glass Bottle | N/A | |||||
15682780
|
Honeywell-Fluka
258105-500ML |
500 mL | Plastic Bottle |
£25.15
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15642780
|
Honeywell-Fluka
258105-1L-PC |
1 L | Glass Bottle | N/A | |||||
15632780
|
Honeywell-Fluka
258105-1L |
1 L | Plastic Bottle |
£32.26
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15652780
|
Honeywell-Fluka
258105-2.5L |
2.5 L | Plastic Bottle |
£64.48
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15628350
|
Honeywell-Fluka
258105-4L |
4 L | Plastic Bottle | N/A | |||||
15672780
|
Honeywell-Fluka
258105-4X4L |
4 x 4 L | Plastic Bottle | N/A | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Specifications
7664-93-9 | |
≤5ppm | |
H2O4S | |
MFCD00064589 | |
2037554 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08g/mol | |
ACS Reagent |
1.840g/mL at 25°C | |
1mmHg (146°C) | |
H2SO4 | |
1830 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
95 to 98% | |
Sulfuric Acid |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H290-H314
missing translation for 'einecsNumber' : 231-639-5
missing translation for 'rtecsNumber' : WS5600000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sulfuric acid
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid