Sulfuric acid, for analysis, ca. 96% solution in water, Thermo Scientific Chemicals
Product Code.: 10624792
Product Code. 10624792
500 mL , Glass Bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code.
10624792
Brand: Thermo Scientific Acros
124645000
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Chemical Identifiers
CAS
7664-93-9
Molecular Formula
H2O4S
Molecular Weight (g/mol)
98.07
MDL Number
MFCD00064589
InChI Key
QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID
1118
ChEBI
CHEBI:26836
IUPAC Name
sulfuric acid
SMILES
OS(O)(=O)=O
Specifications
Melting Point
10.0°C
Color
Colorless
Density
1.8300g/mL
Boiling Point
290.0°C
Ignition Residue
0.0005% max.
Packaging
Glass Bottle
Linear Formula
H2 SO4
Quantity
500 mL
Fieser
01,470; 05,633; 06,558; 07,347; 09,441
Merck Index
15,9104
Specific Gravity
1.83
Solubility Information
Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
Viscosity
21 mPa.s (25°C)
Formula Weight
98.08
Percent Purity
95 to 97%
Grade
Analysis
Physical Form
Liquid
Chemical Name or Material
Sulfuric acid
Show More
Show Less
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number
Certificate Type
Certificate of Analysis
Certificate of Compliance
Certificate of Sterility/Irradiation
Certificate of Validation
Certificate of Guarantee
Certificate of Animal Origin
Certificate of Quality
Certificate of Processing
Declaration of Conformity
Search
5 results found
Lot Number
Certificate Type
Date
Catalog Number
Lot Number A0456861
Certificate Type Certificate of Analysis
Date 10/04/2025
Catalog Number 124640011,124640010,124640100,124640026,124640025,124645001,124645000
Lot Number A0459362
Certificate Type Certificate of Analysis
Date 16/01/2024
Catalog Number 124640011,124640010,124640100,124640026,124640025,124645001,124645000
Lot Number A0456861
Certificate Type Certificate of Analysis
Date 11/10/2023
Catalog Number 124640011,124640010,124640100,124640026,124640025,124645001,124645000
Lot Number A0439907
Certificate Type Certificate of Analysis
Date 11/09/2023
Catalog Number 124640011,124640010,124640100,124640026,124640025,124645001,124645000
Lot Number A0400203
Certificate Type Certificate of Analysis
Date 15/12/2022
Catalog Number 124640011,124640010,124640100,124640026,124640025,124645001,124645000
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sulfuric acid, for analysis, ca. 96% solution in water, Thermo Scientific Chemicals >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.