missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric acid, for analysis, ca. 96% solution in water, Thermo Scientific Chemicals Product Code.: 11904091

Sulfuric acid, for analysis, ca. 96% solution in water, Thermo Scientific Chemicals

Product Code. 11904091
10 L, Plastic drum
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11904091

Brand: Thermo Scientific Acros 124640100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oilShow MoreShow Less
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Melting Point 10.0°C
Color Colorless
Density 1.8300g/mL
Boiling Point 290.0°C
Ignition Residue 0.0005% max.
Packaging Plastic drum
Linear Formula H2SO4
Quantity 10 L
Fieser 01,470; 05,633; 06,558; 07,347; 09,441
Merck Index 15,9104
Specific Gravity 1.83
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
Viscosity 21 mPa.s (25°C)
Formula Weight 98.08
Percent Purity 95 to 97%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Sulfuric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0456861 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number124640011,124640010,124640100,124640026,124640025,124645001,124645000
Lot NumberA0459362 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog Number124640011,124640010,124640100,124640026,124640025,124645001,124645000
Lot NumberA0456861 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog Number124640011,124640010,124640100,124640026,124640025,124645001,124645000
Lot NumberA0439907 Certificate TypeCertificate of Analysis Date11/09/2023 Catalog Number124640011,124640010,124640100,124640026,124640025,124645001,124645000
Lot NumberA0400203 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number124640011,124640010,124640100,124640026,124640025,124645001,124645000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric acid, for analysis, ca. 96% solution in water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.